missing translation for 'onlineSavingsMsg'
Learn More

IKA™ PTFE Coated Stirring Bar Remover

Product Code. 10321352 Shop All IKA Products
Change view
Click to view available options
Length (Metric):
350 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10321352 350 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10321352 Supplier IKA™ Supplier No. 0001293100
  • ONLINE EXCLUSIVE PRICE
    £91.60
    £90.00 / Pack of 1
    Save £1.60
    1% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Description PTFE coated
For Use With (Application) For IKAFLON and TRIKA Stirring Bars Up to 80mm in Length
Diameter (Metric) 8 mm
Length (Metric) 350 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.