missing translation for 'onlineSavingsMsg'
Learn More

JG Finneran Associates™ PTFE/Butyl Rubber Preassembled Screw Caps

Product Code. 12493000
Click to view available options
Color:
Black
Green
Natural
Yellow
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 12493000

Brand: JG Finneran Associates™ 5394F09

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Color Black
Material PTFE, Butyl Rubber
Septum Yes
For Use With (Application) Designed to work in Agilent or other robotic arm autosampler
Includes 9mm red septa
Quantity 1000/Pk.
Type Screw Cap
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.