missing translation for 'onlineSavingsMsg'
Learn More

Cowie Technology™ PTFE Screw-top Bottle

Product Code. 10167941 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
100 mL
150 mL
500 mL
1000 mL
2000 mL
Diameter (Metric) Inner Neck:
10mm
12mm
19mm
25mm
35mm
52mm
57mm
60mm
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Diameter (Metric) Inner Neck unitSize
10167941 1000 mL 57mm Each
10277950 5 mL 10mm Each
10074340 10 mL 12mm Each
10094360 25 mL 19mm Each
10598242 50 mL 25mm Each
10598432 100 mL 35mm Each
10004370 150 mL 35mm Each
10769182 500 mL 52mm Each
10466481 2000 mL 60mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10167941
Only null left
Add to basket
Add to basket

Molded-in hand grips provide ease of manipulation. The PTFE screw-top bottle is chemically resistant and inert.

  • Isostatically molded in pure PTFE and with super smooth internal finish, FDA approved
  • Insoluble and non-contaminating
  • Exceptional thermal resistance can be used from -200°C to +280°C
  • Hand grips for ease of manipulation
  • Do not use, when sealed, for chemical reactions or digestions, loosen lid when thermal sterilizing

Specifications

Capacity (Metric) 1000 mL
Color White
Diameter (Metric) Inner Neck 57mm
For Use With (Application) Storage of hazardous chemicals
Height (Metric) With Closure 185mm
Material PTFE
Max. Temperature (Metric) 280°C
Temperature Range (Metric) -200 to +280°C
Thickness (Metric) 2.75mm
Weight (Metric) 560g
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Closure Included Yes
Graduated No
Product Type Storage Bottle
Diameter (Metric) 100 mm
Closure Type Screw Cap
Autoclavable Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.