Learn More
Bohlender™ PTFE Blunt Tip Tweezers
Use these tweezers for a variety of applications. BOLA PTFE Tweezers feature universal chemical resistance.
Brand: Bohlender™ H912-04
Additional Details : Weight : 0.10000kg
Description
- Precast tweezers
- PTFE construction
- With pointed or blunt ends
- Universal chemical resistance
Specifications
Blunt | |
Precast tweezer | |
150 mm |
Each | |
PTFE | |
Tweezers |