Learn More
Fisherbrand™ PTFE Beaker with Spout
Includes a machined base for good heat transfer. The PTFE beaker includes a molded-in spout for easy pouring.
Brand:  Fisherbrand™ 007.010
Product Code. 10166570
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Non-contaminating and inert to highly corrosive reagents.
  • Non-wettable, lightweight, and heat resistant up to 280°C
  • Machined base for good heat transfer
  • Easy to clean
  • Molded-in spout for easy pouring
  • Isostatically molded from pure PTFE