Learn More
Fisherbrand™ PTFE Beaker Cover
Rest on top of beaker to prevent contamination. The PTFE Beaker Cover is available in a wide range of sizes.
Brand:  Fisherbrand™ 013.075
Product Code. 10379880
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Inert, chemically resistant
  • Smooth internal finish
  • Exceptional thermal resistance: can be used down to -200°C and up to 280°C
  • Isostatically molded from pure PTFE