missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ PTFE Beaker

Product Code. 10122811
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10122811 500 mL Each
10624242 50 mL Each
10081081 100 mL Each
10366111 250 mL Each
10081091 1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10122811 Supplier Kartell™ Supplier No. 0072100
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Possess excellent chemical, mechanical and heat resistance. Kartell™ PTFE Beaker is a non-contaminating beaker that is completely inert to highly corrosive reagents.

  • Very high chemical resistance to acids, alkali and solvents
  • Will withstand temperatures continuously up to 250°C and for short intervals up to 300°C
  • Shockproof, tough and flexible construction with outstanding non-stick properties
  • Suitable for foodstuffs
  • Tall form with spout. Not graduated

Specifications

Color White
Diameter (Metric) Outer 80.5mm
Capacity (Metric) 500 mL
Material PTFE
For Use With (Application) Suitable for Foodstuff
Height (Metric) 122.39mm
Temperature (Metric) Max. Operating 250°C
Type Beaker
Graduated No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.