missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTF1A Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-93539
This item is not returnable.
View return policy
Description
PTF1A Polyclonal specifically detects PTF1A in Zebrafish samples. It is validated for Peptide ELISA.
Specifications
| PTF1A | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| bHLH transcription factor p48, BHLHA29, bHLHa29exocrine pancreas-specific transcription factor p48, Class A basic helix-loop-helix protein 29, p48 DNA-binding subunit of transcription factor PTF1, pancreas specific transcription factor, 1a, pancreas transcription factor 1 subunit alpha, Pancreas-specific transcription factor 1a, PTF1P48, PTF1-p48class II bHLH protein PTF1A | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Zebrafish | |
| IgG |
| ELISA | |
| 0.5 mg/mL | |
| Peptide ELISA Detection limit 1:16000 | |
| NP_997524.1 | |
| PTF1A | |
| Peptide with sequence C-EDPRKLHLKSSINN corresponding to internal region according to Uniprot Zebrafish NP_997524.1. | |
| 0.1 mg | |
| Protein Kinase | |
| 256297 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction