missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PTER Polyclonal Antibody
GREENER_CHOICE

Product Code. 12808902
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12808902 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12808902 Supplier Invitrogen™ Supplier No. PA520750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is human kidney tissue lysate. PA5-20750 can be used with blocking peptide PEP-0864.

PTER is a mammalian homolog to bacterial phosphotriesterases, enzymes that hydrolyze phosphotriester-containing organophosphate pesticides. It is expressed primarily in the proximal renal tubules and the gene has been localized in humans to chromosomal band 10p12 by in situ hybridization. PTER, in addition to FTO, MC4R, and NPC1 has recently been shown to be a new risk loci for early-onset and morbid adult obesity in European populations. At least two isoforms of PTER are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTER
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene PTER
Gene Accession No. Q60866, Q63530, Q96BW5
Gene Alias AI790318; HPHRP; Mpr56-1; Parathion hydrolase-related protein; phosphotriesterase related; phosphotriesterase-related protein; Pter; resiniferatoxin-binding phosphotriesterase-related protein; resiniferatoxin-binding, phosphotriesterase-related; resiniferatoxin-binding, phosphotriesterase-related protein; resiniferotoxin-binding phosphotriesterase-related protein; Rpr1; Rpr-1
Gene Symbols PTER
Host Species Rabbit
Immunogen A 16 amino acid peptide from near the carboxy terminus of human PTER.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19212, 63852, 9317
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.