missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PTEN2/TPTE Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
Specifications
Specifications
| Gene ID (Entrez) | 7179 |
| Species | Human |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-41395 |
| Gene Alias | Cancer/testis antigen 44, EC 3.1.3.48, PTEN2, putative tyrosine-protein phosphatase TPTE, tensin, putative protein-tyrosine phosphatase, transmembrane phosphatase with tensin homologytensin, putative protein-tyrosine phosphatase, EC 3.1.3.4810 CT44PTEN-related tyrosine phosphatase, Tumor antigen BJ-HCC-5 |
| Gene Symbol | TPTE |
| Quantity | 0.1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction