missing translation for 'onlineSavingsMsg'
Learn More

PTEN2/TPTE Antibody, Novus Biologicals™

Product Code. 18354188 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18354188 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18354188 Supplier Novus Biologicals Supplier No. H00007179B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

PTEN2/TPTE Polyclonal antibody specifically detects PTEN2/TPTE in Human samples. It is validated for Western Blot, Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTEN2/TPTE
Applications Western Blot, Flow Cytometry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS (pH 7.4)
Gene Accession No. AAH28719.1
Gene Alias Cancer/testis antigen 44, EC 3.1.3.48, PTEN2, putative tyrosine-protein phosphatase TPTE, tensin, putative protein-tyrosine phosphatase, transmembrane phosphatase with tensin homologytensin, putative protein-tyrosine phosphatase, EC 3.1.3.4810CT44PTEN-related tyrosine phosphatase, Tumor antigen BJ-HCC-5
Host Species Mouse
Immunogen TPTE (AAH28719.1, 1 a.a. - 533 a.a.) full-length human protein. MNESPDPTDLAGVIIELGPNDSPQTSEFKGATEEAPAKESVLARLSKFEVEDAENVASYDSKIKKIVHSIVSSFAFGLFGVFLVLLDVTLILADLIFTDSKLYIPLEYRSISLAIALFFLMDVLLRVFVERRQQYFSDLFNILDTAIIVILLLVDVVYIFFDIKLLRNIPRWTHLLRLLRLIILLRIFHLFHQKRQLEKLIRRRVSENKRRYTRDGFDLDLTYVTERIIAMSFPSSGRQSFYRNPIKEVVRFLDKKHRNHYRVYNLCSERAYDPKHFHNRVVRIMIDDHNVPTLHQMVVFTKEVNEWMAQDLENIVAIHCKGGTDRTGTMVCAFLIASEICSTAKESLYYFGERRTDKTHSEKFQGVETPSQKRYVAYFAQVKHLYNWNLPPRRILFIKHFIIYSIPRYVRDLKIQIEMEKKVVFSTISLGKCSVLDNITTDKILIDVFDGPPLYDDVKVQFFYSNLPTYYDNCSFYFWLHTSFIENNRLYLPKNELDNLHKQKARRIYPSDFAVEILFGEKMTSSDVVAGSD
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7179
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.