missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTEN2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113641
This item is not returnable.
View return policy
Description
Immunogen sequence: YFAQVKHLYN WNLPPRRILF IKHFIIYSIP RYVRDLKIQI EMEKKVVFST For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%.
TPTE encodes a putative transmembrane tyrosine phosphatase that may be involved in signal transduction pathways of the endocrine or spermatogenetic function of the testis.
Specifications
| PTEN2 | |
| Polyclonal | |
| Unconjugated | |
| TPTE | |
| Cancer/testis antigen 44; CT44; Pten2; PTEN-related tyrosine phosphatase; Putative tyrosine-protein phosphatase TPTE; tensin, putative protein-tyrosine phosphatase; TPTE; Transmembrane phosphatase with tensin homology; tumor antigen BJ-HCC-5 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 7179 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P56180 | |
| TPTE | |
| Synthetic peptide directed towards the middle region of human TPTE. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido