missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTEN Monoclonal Antibody (17.A), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA512278
This item is not returnable.
View return policy
Description
MA5-12278 targets PTEN in WB, IHC and ICC/IF applications and shows reactivity with Human and Mouse samples. The MA5-12278 immunogen is recombinant human PTEN protein.
PTEN was identified as a tumor suppressor that is mutated in a large number of cancers at high frequency. The PTEN gene is a phosphatidylinositol-3, 4, 5-trisphosphate 3-phosphatase. It contains a tensin like domain as well as a catalytic domain similar to that of the dual specificity protein tyrosine phosphatases. Unlike most of the protein tyrosine phosphatases, this protein preferentially dephosphorylates phosphoinositide substrates. PTEN negatively regulates intracellular levels of phosphatidylinositol-3, 4, 5-trisphosphate in cells and functions as a tumor suppressor by negatively regulating AKT/PKB signaling pathway. PTEN plays a role as a key modulator of the AKT-mTOR signaling pathway controlling the tempo of the process of newborn neurons integration during adult neurogenesis, including correct neuron positioning, dendritic development and synapse formation. Also, PTEN may be a negative regulator of insulin signaling and glucose metabolism in adipose tissue. The nuclear monoubiquitinated form possesses greater apoptotic potential, whereas the cytoplasmic nonubiquitinated form of PTEN induces less tumor suppressive ability.
Specifications
| PTEN | |
| Monoclonal | |
| Conc. Not Determined | |
| tissue culture supernatant with 0.09% sodium azide | |
| O08586, P60484 | |
| PTEN | |
| Recombinant human PTEN protein | |
| RUO | |
| 19211, 5728 | |
| 4° C | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 17.A | |
| Unconjugated | |
| PTEN | |
| 10q23del; 2310035O07Rik; A130070J02Rik; AI463227; B430203M17Rik; BZS; CWS1; DEC; GLM2; homolog of human mutated in multiple advanced cancers; MHAM; mitochondrial phosphatase and tensin protein alpha; mitochondrial PTENalpha; Mmac; MMAC1; MMAC1 phosphatase and tensin homolog deleted on chromosome 10; mutated in multiple advanced cancers 1; Phosphatase and tensin homolog; phosphatase and tensin homolog (mutated in multiple advanced cancers 1); phosphatase and tensin homolog deleted on chromosome ten; phosphatase and tensin-like protein; phosphatidylinositol 3; phosphatidylinositol 3,4,5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN; phosphatidylinositol-3; phosphatidylinositol-3,4,5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN; protein tyrosine phosphatase and tensin-like protein; PTEN; pten tumor suppressor; PTEN1; TEP1 | |
| Mouse | |
| 500 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgM κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction