missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTDSS2 Recombinant Rabbit Monoclonal Antibody (K01_1X30)
Description
PTDSS2 Recombinant Monoclonal Antibody for Western Blot
Catalyzes a base-exchange reaction in which the polar head group of phosphatidylethanolamine (PE) or phosphatidylcholine (PC) is replaced by L-serine. PTDSS2 is specific for phosphatatidylethanolamine and does not act on phosphatidylcholine.
Specifications
Specifications
| Antigen | PTDSS2 |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_1X30 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4 |
| Gene | PTDSS2 |
| Gene Accession No. | Q9BVG9 |
| Gene Alias | phosphatidylserine synthase 2; PSS2; PSS-2; ptdSer synthase 2; PTDSS2; Serine-exchange enzyme II |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?