missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTDSS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553505
This item is not returnable.
View return policy
Description
Immunogen sequence: AEHYGHREKT YSECEDGTYS PEISWHHRKG TKGSEDSPPK HAGNNESHSS RRRNRHSKSK VTNGVGKK Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 88%.
Catalyzes a base-exchange reaction in which the polar head group of phosphatidylethanolamine (PE) or phosphatidylcholine (PC) is replaced by L-serine. In membranes, PTDSS1 catalyzes mainly the conversion of phosphatidylcholine. Also converts, in vitro and to a lesser extent, phosphatidylethanolamine.
Specifications
| PTDSS1 | |
| Polyclonal | |
| Unconjugated | |
| PTDSS1 | |
| KIAA0024; LMHD; Phosphatidylserine synthase 1; PSS1; PSS-1; PSSA; ptdSer synthase 1; PTDSS1; Serine-exchange enzyme I | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9791 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P48651 | |
| PTDSS1 | |
| Recombinant protein corresponding to Human PTDSS1. Recombinant protein control fragment (Product #RP-92424). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction