missing translation for 'onlineSavingsMsg'
Learn More

PTCRA Antibody - BSA Free, Novus Biologicals™

Product Code. 18668110 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.01mL
0.02mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18668110 0.02 mL 0.02mL
18672661 0.1 mL 0.01mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18668110 Supplier Novus Biologicals Supplier No. NBP2930050.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PTCRA Polyclonal antibody specifically detects PTCRA in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTCRA
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias pre T-cell antigen receptor alpha, PTA, PT-ALPHA
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 172-281 of human PTCRA (NP_612153.2). LCDPAGPLPSPATTTRLRALGSHRLHPATETGGREATSSPRPQPRDRRWGDTPPGRKPGSPVWGEGSYLSSYPTCPAQAWCSRSALRAPSSSLGAFFAGDLPPPLQAGAA
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 171558
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.