missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTCHD2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572748
This item is not returnable.
View return policy
Description
PTCHD2 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Plays a role in neuronal proliferation and differentiation (PubMed:25281927). Plays a role in the accumulation of cellular cholesterol. Involved in intracellular lipid droplet formation (PubMed:25281927). May contribute to cholesterol homeostasis in neuronal cells. [UniProt].
Specifications
| PTCHD2 | |
| Polyclonal | |
| Unconjugated | |
| DISP3 | |
| DISP3; dispatched 3; dispatched homolog 3; dispatched RND transporter family member 3; G630052C06Rik; Kiaa1337; patched domain containing 2; patched domain-containing protein 2; Protein dispatched homolog 3; Ptchd2; RGD1305820; RNDEu-2; RND-type protein RNDEu-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 242748, 313705, 57540 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| A3KFU9, Q9P2K9 | |
| DISP3 | |
| PTCHD2 antibody for a 19 amino acid synthetic peptide from near the center of human PTCHD2. The immunogen is within amino acids 780 - 830 of PTCHD2. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction