missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTCH1, Mouse anti-Human, Clone: 1G2, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00005727-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: PKMWLHYFRDWLQGLQDAFDSDWETGKIMPNNYKNGSDDGVLAYKLLVQTGSRDKPIDISQLTKQRLVDADGIINPSAFYIYLTAWVSNDPVAYAASQANSpecifications
| PTCH1 | |
| Monoclonal | |
| Unconjugated | |
| patched homolog 1 (Drosophila) | |
| BCNS/FLJ26746/FLJ42602/HPE7/NBCCS/PTC/PTC1/PTCH/PTCH11 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1G2 | |
| In 1x PBS, pH 7.4 | |
| NM_000264 | |
| PTCH1 | |
| PTCH (NP_000255, 841 a.a. ∼ 940 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 5727 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction