missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTCH Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA146222
This item is not returnable.
View return policy
Description
Suggested positive control: antigen standard for PTCH1 (transient overexpression lysate). Reacts with human, mouse, and rat.
This gene encodes a member of the patched gene family. The encoded protein is the receptor for sonic hedgehog, a secreted molecule implicated in the formation of embryonic structures and in tumorigenesis, as well as the desert hedgehog and indian hedgehog proteins. This gene functions as a tumor suppressor. Mutations of this gene have been associated with basal cell nevus syndrome, esophageal squamous cell carcinoma, trichoepitheliomas, transitional cell carcinomas of the bladder, as well as holoprosencephaly. Alternative splicing results in multiple transcript variants encoding different isoforms. Additional splice variants have been described, but their full length sequences and biological validity cannot be determined currently.
Specifications
| PTCH | |
| Polyclonal | |
| Unconjugated | |
| Ptch1 | |
| A230106A15Rik; BCNS; etID309849.2; FLJ26746; FLJ42602; HGNC:9585; HPE7; lep; leprechaun; mes; mesenchymal dysplasia; NBCCS; OTTHUMP00000021709; OTTHUMP00000021710; patched; Patched 1; patched 2; patched homolog 1; patched homolog 2; patched protein; patched1; patched-2 protein; patched-like protein; protein patched homolog 1; Ptc; ptc1; ptc-1; ptc2; Ptch; PTCH protein +12b; PTCH protein +4'; PTCH protein -10; PTCH protein -3; PTCH protein -3,4,5; PTCH1; Ptch1 protein +12b; PTCH11; Ptch2; RP11-435O5.3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 19206, 5727, 89830 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q13635, Q61115 | |
| Ptch1 | |
| A synthetic peptide made to residues 269-279 of the human and mouse PTCH protein. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction