missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTCH Polyclonal Antibody

PTCH Polyclonal Antibody
Brand: Invitrogen PA5110235
This item is not returnable.
View return policy
Description
PTCH Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| PTCH | |
| Polyclonal | |
| Unconjugated | |
| Ptch1 | |
| A230106A15Rik; BCNS; etID309849.2; FLJ26746; FLJ42602; HGNC:9585; HPE7; lep; leprechaun; mes; mesenchymal dysplasia; NBCCS; OTTHUMP00000021709; OTTHUMP00000021710; patched; Patched 1; patched 2; patched homolog 1; patched homolog 2; patched protein; patched1; patched-2 protein; patched-like protein; protein patched homolog 1; Ptc; ptc1; ptc-1; ptc2; Ptch; PTCH protein +12b; PTCH protein +4'; PTCH protein -10; PTCH protein -3; PTCH protein -3,4,5; PTCH1; Ptch1 protein +12b; PTCH11; Ptch2; RP11-435O5.3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 19206, 5727, 89830 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 2.77 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q13635, Q61115 | |
| Ptch1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 770-970 of human PTCH1 (NP_000255.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction