missing translation for 'onlineSavingsMsg'
Learn More

PTC-124 Sodium Salt, TRC

Product Code. 30396700
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396700 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30396700 Supplier Toronto Research Chemicals Supplier No. TRCF5960701G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material Ataluren Sodium
CAS 775304-59-1
Molecular Formula C15H8FN2NaO3
Quantity 1 g
Synonym 3-[5-(2-Fluorophenyl)-1,2,4-oxadiazol-3-yl]benzoic Acid Sodium Salt
InChI Formula InChI=1S/C15H9FN2O3.Na/c16-12-7-2-1-6-11(12)14-17-13(18-21-14)9-4-3-5-10(8-9)15(19)20;/h1-8H,(H,19,20);/q;+1/p-1
SMILES FC1=CC=CC=C1C2=NC(C3=CC(C(O[Na])=O)=CC=C3)=NO2
IUPAC Name sodium 3-(5-(2-fluorophenyl)-1,2,4-oxadiazol-3-yl)benzoate
Molecular Weight (g/mol) 306.22
Formula Weight 306.04
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.