missing translation for 'onlineSavingsMsg'
Learn More

PTC-028, MedChemExpress

Product Code. 16460257
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16460257 50 mg 50mg
16456617 5 mg 5mg
16452896 25 mg 25mg
16423898 10 mg 10mg
16414936 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16460257 Supplier Medchem Express Supplier No. HY103696

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PTC-028 is an orally bioavailable inhibitor of stem cell factor BMI-1 in ovarian cancer. PTC-028 selectively inhibits cancer cells whereas normal cells remain unaffected. PTC-028 downregulates BMI-1, inducing caspase-mediated apoptosis.

PTC-028 is an orally bioavailable inhibitor of stem cell factor BMI-1 in ovarian cancer. PTC-028 selectively inhibits cancer cells whereas normal cells remain unaffected. PTC-028 downregulates BMI-1, inducing caspase-mediated apoptosis.

Specifications

Chemical Name or Material PTC-028
CAS 1782970-28-8
Color White
Purity Grade Notes Research
Molecular Formula C19H12F5N5
Quantity 50 mg
Solubility Information DMSO : 125 mg/mL (308.40 mM; Need ultrasonic)
SMILES FC(C1=CC=C(NC2=NC(N3C4=CC(F)=C(F)C=C4N=C3C)=CN=C2)C=C1)(F)F
Molecular Weight (g/mol) 405.32
Formula Weight 405.32
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.