missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTBP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569084
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 79%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
This gene is apparently required for both cerebellar and cortical development in humans. This gene mutations cause specific forms of Joubert syndrome-related disorders. Joubert syndrome (JS) is a recessively inherited developmental brain disorder with several identified causative chromosomal loci. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| PTBP3 | |
| Polyclonal | |
| Unconjugated | |
| PTBP3 | |
| 5830471K22Rik; AA407443; AI462022; AW107884; C86549; fission yeast differentiation regulator; polypyrimidine tract binding protein 3; polypyrimidine tract-binding protein 3; PTBP3; regulator of differentiation (in S. pombe) 1; regulator of differentiation 1; regulator of differentiation 1; polypyrimidine tract-binding protein 3; ROD1; ROD1 regulator of differentiation 1; si:dkey-27p1.2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 230257 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8BHD7 | |
| PTBP3 | |
| synthetic peptide corresponding to a region of mouse PTBP3 (aa 84-133). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction