missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTBP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101026
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PTBP2.
PTBP2 encodes a protein that binds to the intronic cluster of RNA regulatory elements, downstream control sequence (DCS). It is implicated in controlling the assembly of other splicing-regulatory proteins. This protein is very similar to the polypyrimidine tract binding protein but it is expressed primarily in the brain.
Specifications
| PTBP2 | |
| Polyclonal | |
| Unconjugated | |
| PTBP2 | |
| br n nPTB5; brain-enriched polypyrimidine tract-binding protein; Brain-enriched PTB; Brptb; neural polypyrimidine tract binding protein; neural polypyrimidine tract-binding protein; Neurally-enriched homolog of PTB; NPTB; nPTB5; nPTB6; nPTB7; nPTB8; polypyrimidine tract binding protein 2; polypyrimidine tract-binding protein 2; PTB; Ptb2; PTB-like protein; Ptblp; PTBP2; RRM-type RNA-binding protein brPTB | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 310820, 56195, 58155 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q66H20, Q91Z31, Q9UKA9 | |
| PTBP2 | |
| A synthesized peptide derived from human PTBP2(Accession Q9UKA9), corresponding to amino acid residues K256-A306. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction