missing translation for 'onlineSavingsMsg'
Learn More

PTBP1 Antibody (3H8), Novus Biologicals™

Product Code. 18378968 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18378968 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18378968 Supplier Novus Biologicals Supplier No. H00005725M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PTBP1 Monoclonal antibody specifically detects PTBP1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Single Cell Western
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTBP1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Single Cell Western
Classification Monoclonal
Clone 3H8
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry 1:10 to 1:500, Immunocytochemistry/ Immunofluorescence 1:10 to 1:500, Immunoprecipitation 1:10 to 1:500, Immunohistochemistry-Paraffin 1:10 to 1:500, Single Cell Western 1:10
Formulation PBS (pH 7.4)
Gene Accession No. NP_002810
Gene Alias Heterogeneous nuclear ribonucleoprotein I, heterogeneous nuclear ribonucleoprotein polypeptide I, hnRNP I, HNRNPI, HNRNP-I, HNRPI, MGC10830, MGC8461, polypyrimidine tract binding protein (heterogeneous nuclear ribonucleoproteinI), polypyrimidine tract binding protein 1, polypyrimidine tract-binding protein 1, pPTB, PTB-1, PTB2, PTB3, PTB4, PTB57 kDa RNA-binding protein PPTB-1, PTB-T, RNA-binding protein
Host Species Mouse
Immunogen PTBP1 (NP_002810, 45 a.a. ~ 144 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. KKFKGDSRSAGVPSRVIHIRKLPIDVTEGEVISLGLPFGKVTNLLMLKGKNQAFIEMNTEEAANTMVNYYTSVTPVLRGQPIYIQFSNHKELKTDSSPNQ
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5725
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.