missing translation for 'onlineSavingsMsg'
Learn More

Pt(II) meso-Tetra(pentafluorophenyl)porphine, TRC

Product Code. 30418610
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30418610 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30418610 Supplier Toronto Research Chemicals Supplier No. TRCP8401955MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pt(II) meso-Tetra(pentafluorophenyl)porphine
CAS 109781-47-7
Molecular Formula C44H8F20N4Pt
Quantity 5 mg
Synonym Platinum','[5,10,15,20-tetrakis(pentafluorophenyl)-21H,23H-porphinato(2-)-κN21,κN22,κN23,κN24]-','(SP-4-1)-, Platinum','[5,10,15,20-tetrakis(pentafluorophenyl)-21H,23H-porphinato(2-)-N21,N22,N23,N24]-','(SP-4-1)-, (SP-4-1)-[5,10,15,20-Tetrakis(pentafluorophenyl)-21H,23H-porphinato(2-)-κN21,κN22,κN23,κN24]platinum, 21H,23H-Porphine','5,10,15,20-tetrakis(pentafluorophenyl)-','platinum complex, Platinum 5,10,15,20-tetrakis(pentafluorophenyl)porphyrin, Platinum meso-tetra(pentafluorophenyl)porphine, Platinum tetra(pentafluorophenyl)porphyrin, Platinum tetrakis(pentafluorophenyl)porphyrin, Platinum(II) tetra(pentafluorophenyl)porphine, Platinum(II)-5,10,15,20-tetrakis(2,3,4,5,6-pentafluorophenyl)porphyrin, PtTFPP, meso-5,10,15,20-Tetrakis(2,3,4,5,6-pentafluorophenyl)porphyrinatoplatinum, meso-5,10,15,20-Tetrakis(pentafluorophenyl)porphyrinatoplatinum, Pt(II) meso-Tetra(pentafluorophenyl)porphine, Platinum(II) meso-tetra(pentafluorophenyl)porphine
InChI Formula InChI=1S/C44H8F20N4.Pt/c45-25-21(26(46)34(54)41(61)33(25)53)17-9-1-2-10(65-9)18(22-27(47)35(55)42(62)36(56)28(22)48)12-5-6-14(67-12)20(24-31(51)39(59)44(64)40(60)32(24)52)16-8-7-15(68-16)19(13-4-3-11(17)66-13)23-29(49)37(57)43(63)38(58)30(23)50;/h1-8H;/q-2;+2/b17-9+,17-11?,18-10+,18-12?,19-13+,19-15?,20-14+,20-16?;
SMILES Fc1c(F)c(F)c(C2=C3C=CC4=C(C5=N6C(=C(C7=CC=C8C(=C9C=CC2=N9[Pt+2]6([N-]34)[N-]78)c%10c(F)c(F)c(F)c(F)c%10F)c%11c(F)c(F)c(F)c(F)c%11F)C=C5)c%12c(F)c(F)c(F)c(F)c%12F)c(F)c1F
Molecular Weight (g/mol) 1167.61
Formula Weight 1167.0077
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.