Description
- Flexible
- Additional sensor
- Interior
Specifications
Specifications
| Description | Pt 100 temperature sensor - additional flexible Pt 100, interior, for displaying the temperature on the unit display |
| For Use With (Equipment) | BINDER Chambers Series MK |
| Material | Plastic |
| Type | Temperature Sensor |