missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSTPIP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578276
This item is not returnable.
View return policy
Description
Predicted Reactivity: Mouse (83%), Rat (80%), Pig (85%), Bovine (86%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PSTPIP1 is involved in regulation of the actin cytoskeleton. May regulate WAS actin-bundling activity. Bridges the interaction between ABL1 and PTPN18 leading to ABL1 dephosphorylation. May play a role as a scaffold protein between PTPN12 and WAS and allow PTPN12 to dephosphorylate WAS. Has the potential to physically couple CD2 and CD2AP to WAS. Acts downstream of CD2 and CD2AP to recruit WAS to the T-cell:APC contact site so as to promote the actin polymerization required for synapse induction during T-cell activation. Down-regulates CD2-stimulated adhesion through the coupling of PTPN12 to CD2. Also has a role in innate immunity and the inflammatory response. Recruited to inflammasomes by MEFV. Induces formation of pyroptosomes, large supramolecular structures composed of oligomerized PYCARD dimers which form prior to inflammatory apoptosis. Binding to MEFV allows MEFV to bind to PYCARD and facilitates pyroptosome formation. Regulates endocytosis and cell migration in neutrophils.
Specifications
| PSTPIP1 | |
| Polyclonal | |
| Unconjugated | |
| PSTPIP1 | |
| CD2 antigen-binding protein 1; CD2 cytoplasmic tail-binding protein; CD2-binding protein 1; CD2BP1; CD2BP1L; CD2BP1S; def-2; H-PIP; PAPAS; PEST phosphatase-interacting protein 1; proline-serine-threonine phosphatase interacting protein 1; proline-serine-threonine phosphatase-interacting protein 1; PSTPIP; Pstpip1; truncated proline-serine-threonine phosphatase interacting protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9051 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.63 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| O43586 | |
| PSTPIP1 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human PSTPIP1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction