missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSRC2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-68265
This item is not returnable.
View return policy
Description
PSRC2 Polyclonal specifically detects PSRC2 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| PSRC2 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| coiled-coil domain containing 131, Coiled-coil domain-containing protein 131, DKFZp686A0722, KIAA0546CCDC131, MGC23401, MGC90200, proline/serine-rich coiled-coil 2, Proline/serine-rich coiled-coil protein 2, PSRC2, zinc finger C3H1 domain-containing protein, zinc finger, C3H1-type containing | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 196441 | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 1:10-1:500 | |
| NP_659419.3 | |
| ZFC3H1 | |
| The immunogen recognized by this antibody maps to a region between residue 300 and 350 of human coiled-coil domain containing 131 using the numbering given in entry NP_659419.3 (GeneID 196441). | |
| 0.1 mL | |
| Primary | |
| Based on 100% sequence identity, this antibody is predicted to react with Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Duckbill platypus, Tasmanian devil, White-tufted-ear marmoset, Crab-eating macaque and Northern white-cheeked gibbon. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction