missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSPN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550530
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total Persephin protein.
Persephin is a disulfide-linked, homodimeric, neurotrophic factor structurally related to GDNF, artemin, and neurturin. These proteins belong to the cysteine-knot family of growth factors that assume stable dimeric structures. Persephin signals through a multicomponent receptor system, composed of RET and one of four GFR alpha (alpha1-alpha4) receptors. The GFRalpha4 was first identified in chicken, and was later shown to be the preferential binding subunit for persephin. Persephin promotes the survival of ventral midbrain dopaminergic neurons and motor neurons after sciatic nerve oxotomy, and, like GDNF, promotes ureteric bud branching. However, in contrast to GDNF and neurturin, persephin does not support the survival of peripheral neurons.
Specifications
| PSPN | |
| Polyclonal | |
| Unconjugated | |
| PSPN | |
| Persephin; PSP; Pspn | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5623 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.8 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O60542 | |
| PSPN | |
| Synthetic peptide corresponding to a region derived from internal residues of human persephin. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction