missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSPHL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-32723
This item is not returnable.
View return policy
Description
PSPHL Polyclonal specifically detects PSPHL in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| PSPHL | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| PSPHP1 | |
| Full length human PSPH Recombinant protein. | |
| Affinity Purified | |
| RUO | |
| 8781 | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
| CO9, phosphoserine phosphatase pseudogene 1, PSPHL | |
| Rabbit | |
| 25 kDa | |
| 100 μL | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction