missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PSPC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30461
This item is not returnable.
View return policy
Description
PSPC1 Polyclonal antibody specifically detects PSPC1 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| PSPC1 | |
| Polyclonal | |
| Tris-buffered Saline 0.1% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 55269 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| FLJ33554, paraspeckle component 1, paraspeckle protein 1, PSP1 | |
| between 1 and 50 | |
| 20 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction