missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSP94/MSMB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Specifications
| Antigen | Prostate Secretory Protein/PSP |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
PSP94/MSMB Polyclonal specifically detects PSP94/MSMB in Human samples. It is validated for Western Blot.Specifications
| Prostate Secretory Protein/PSP | |
| Polyclonal | |
| Rabbit | |
| Cancer | |
| NP_002434 | |
| 4477 | |
| The immunogen for this antibody is Prostate Secretory Protein/PSP. Peptide sequence ETEISCCTLVSTPVGYDKDNCQRIFKKEDCKYIVVEKKDPKKTCSVSEWI. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| beta-microseminoprotein, IGBFProstate secreted seminal plasma protein, immunoglobulin binding factor, Immunoglobulin-binding factor, microseminoprotein, beta-, MSP, MSPB, PN44Prostate secretory protein of 94 amino acids, prostatic secretory protein 94, PRPS, PRSP, PSP, PSP57, PSP94HPC13, PSP-94Seminal plasma beta-inhibin | |
| MSMB | |
| IgG | |
| 11 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title