missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Psp, Mouse anti-Mouse, Clone: 1F12, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB16358.50ug
This item is not returnable.
View return policy
Specifications
| Psp | |
| Monoclonal | |
| Unconjugated | |
| In PBS (0.05% protein stabilizer, 0.05% sodium azide). | |
| - | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1F12 | |
| ELISA (1:10000) Flow Cytometry (1:200-1:400) Immunocytochemistry (1:200-1:1000) Immunofluorescence (1:1000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
| parotid secretory protein | |
| Psp | |
| Recombinant protein corresponding to mouse Psp from E. coli. | |
| RUO | |
| 19194 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction