missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSP/BPIFA2 Antibody (5B2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37512
This item is not returnable.
View return policy
Description
PSP/BPIFA2 Monoclonal specifically detects PSP/BPIFA2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| PSP/BPIFA2 | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| BPIFA2 | |
| Purified recombinant fragment of human Splunc2 expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| 140683 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA | |
| 5B2 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 | |
| bA49G10.1, chromosome 20 open reading frame 70, Parotid secretory protein, PSP, short palate, lung and nasal epithelium carcinoma associated 2, short palate, lung and nasal epithelium carcinoma-associated protein 2, SPLUNC2 | |
| Mouse | |
| 27 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction