missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSORS1C2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563936
This item is not returnable.
View return policy
Description
Immunogen sequence: SGSEGHPSHP PAEDREEAGS PTLPQG Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 50%.
Specifications
| PSORS1C2 | |
| Polyclonal | |
| Unconjugated | |
| PSORS1C2 | |
| C6orf17; Protein SPR1; psoriasis susceptibility 1 candidate 2; psoriasis susceptibility 1 candidate gene 2 protein; PSORS1C2; SPR1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 170680 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UIG4 | |
| PSORS1C2 | |
| Recombinant protein corresponding to Human PSORS1C2. Recombinant protein control fragment (Product #RP-99608). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction