missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PSNCBAM-1

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
PSNCBAM-1 is a CB1 receptor negative allosteric modulator (IC50 values are 45 and 209 nM for the inhibition of CP 55,940 and WIN 55,212-2 respectively). Has no effect at the CB2 receptor. Decreases food intake and body weight in rats.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | PSNCBAM-1 |
| Purity | >99% |
| Molecular Weight (g/mol) | 392.88 |
| Inhibitors | PSNCBAM-1 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction