missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSMG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531298
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T. Predicted reactivity: Mouse (83%), Rat (81%), Pig (89%), Rabbit (84%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Chaperone protein which promotes assembly of the 20S proteasome as part of a heterodimer with PSMG2. The PSMG1-PSMG2 heterodimer binds to the PSMA5 and PSMA7 proteasome subunits, promotes assembly of the proteasome alpha subunits into the heteroheptameric alpha ring and prevents alpha ring dimerization.
Specifications
| PSMG1 | |
| Polyclonal | |
| Unconjugated | |
| PSMG1 | |
| AW552102; C21LRP; C21-LRP; Chromosome 21 leucine-rich protein; Down syndrome critical region gene 2; Down syndrome critical region homolog 2; Down syndrome critical region protein 2; Down syndrome critical region protein 2 homolog; Dscr2; leucine rich protein C21-LRP; LRPC21; PAC1; PAC-1; proteasome (prosome, macropain) assembly chaperone 1; proteasome assembling chaperone 1; proteasome assembly chaperone 1; Psmg1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8624 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.91 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| O95456 | |
| PSMG1 | |
| Recombinant protein encompassing a sequence within the center region of human PSMG1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction