missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSMF1 Monoclonal Antibody (OTI2D7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526099
This item is not returnable.
View return policy
Description
PSMF1 Monoclonal Antibody for Western Blot, ICC/IF
The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a protein that inhibits the activation of the proteasome by the 11S and 19S regulators. Alternative transcript variants have been identified for this gene.
Specifications
| PSMF1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q92530 | |
| PSMF1 | |
| Full length human recombinant protein of PSMF1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot, Immunocytochemistry | |
| OTI2D7 | |
| Unconjugated | |
| PSMF1 | |
| AW048666; BC012260; hPI31; PI31; proteasome (prosome, macropain) inhibitor subunit 1; proteasome (prosome, macropain) inhibitor subunit 1 (PI31); proteasome inhibitor PI31 subunit; proteasome inhibitor subunit 1; PSMB2; Psmf1; RP23 402M7.5; RP4 545L17.1; RP4-545L17.1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9491 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction