missing translation for 'onlineSavingsMsg'
Learn More

PSMF1 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18336792 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18336792 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18336792 Supplier Novus Biologicals Supplier No. NBP299618JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PSMF1 Polyclonal specifically detects PSMF1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMF1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias hPI31, PI31, proteasome (prosome, macropain) inhibitor subunit 1 (PI31), proteasome inhibitor hP131 subunit, proteasome inhibitor PI31 subunit
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human PSMF1.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 9491
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.