missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSME4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94690-0.02ml
This item is not returnable.
View return policy
Description
PSME4 Polyclonal antibody specifically detects PSME4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PSME4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| proteasome (prosome, macropain) activator subunit 4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1634-1843 of human PSME4 (NP_055429.2). LYPHQVPLVLQVLKQTARSSSWHARYTVLTYLQTMVFYNLFIFLNNEDAVKDIRWLVISLLEDEQLEVREMAATTLSGLLQCNFLTMDSPMQIHFEQLCKTKLPKKRKRDPGSVGDTIPSAELVKRHAGVLGLGACVLSSPYDVPTWMPQLLMNLSAHLNDPQPIEMTVKKTLSNFRRTHHDNWQEHKQQFTDDQLLVLTDLLVSPCYYA | |
| 0.02 mL | |
| Neuroscience, Ubiquitin Proteasome Pathway | |
| 23198 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction