Learn More
PSME3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14907-1-AP-150UL
 View more versions of this product
Product Code. 16813814
  • £349.00 / 150µL
Description
SFRS10 contains 1 RRM (RNA recognition motif) domain and belongs to the splicing factor SR family. It is a sequence-specific RNA-binding protein which participates in the control of pre-mRNA splicing.
Specifications
Specifications
PSME3
Polyclonal
Unconjugated
PSME3
Ki, Ki nuclear autoantigen, PA28 gamma, PA28G, PA28gamma, PSME3, REG GAMMA
Rabbit
Antigen Affinity Chromatography
RUO
10197, 19192, 287716
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P61289, P61290
Psme3
PSME3 Fusion Protein Ag6702
150 μL
Primary
Human, Mouse, Rat