missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSME3 Polyclonal Antibody

PSME3 Polyclonal Antibody
Brand: Invitrogen PA580952
This item is not returnable.
View return policy
Description
PSME3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP, ELISA
Specifications
| PSME3 | |
| Polyclonal | |
| Unconjugated | |
| PSME3 | |
| 11S regulator complex gamma subunit; 11S regulator complex subunit gamma; AA410043; Ab2-371; Activator of multicatalytic protease subunit 3; AU020960; epididymis secretory protein Li 283; HEL-S-283; Ki; Ki antigen; Ki nuclear autoantigen; PA28 gamma variant 5; pa28g; PA28gamma; PA28-gamma; prosome macropain; proteaseome (prosome, macropain) 28 subunit, 3; proteaseome (prosome, macropain) activator subunit 3 (PA28 gamma, Ki); proteasome (prosome, macropain) activator subunit 3; proteasome (prosome, macropain) activator subunit 3 (PA28 gamma; Ki); proteasome activator 28 subunit gamma; proteasome activator 28-gamma; proteasome activator complex subunit 3; proteasome activator subunit 3; Psme3; REG gamma-3; REGgamma; REG-gamma | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 10197 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with no preservative | |
| P61289 | |
| PSME3 | |
| Recombinant Human PSME3 Protein (Met1-Tyr254). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction