missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSME3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ 383800
This item is not returnable.
View return policy
Description
PSME3 Polyclonal Antibody for Western Blot, IP
SFRS10 contains 1 RRM (RNA recognition motif) domain and belongs to the splicing factor SR family. It is a sequence-specific RNA-binding protein which participates in the control of pre-mRNA splicing.
Specifications
| PSME3 | |
| Polyclonal | |
| Unconjugated | |
| PSME3 | |
| 11S regulator complex gamma subunit; 11S regulator complex subunit gamma; AA410043; Ab2-371; Activator of multicatalytic protease subunit 3; AU020960; epididymis secretory protein Li 283; HEL-S-283; Ki; Ki antigen; Ki nuclear autoantigen; PA28 gamma variant 5; pa28g; PA28gamma; PA28-gamma; prosome macropain; proteaseome (prosome, macropain) 28 subunit, 3; proteaseome (prosome, macropain) activator subunit 3 (PA28 gamma, Ki); proteasome (prosome, macropain) activator subunit 3; proteasome (prosome, macropain) activator subunit 3 (PA28 gamma; Ki); proteasome activator 28 subunit gamma; proteasome activator 28-gamma; proteasome activator complex subunit 3; proteasome activator subunit 3; Psme3; REG gamma-3; REGgamma; REG-gamma | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10197, 19192, 287716 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.25 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| P61289, P61290 | |
| PSME3 | |
| Synthetic peptide derived from the N-terminal region of the human, mouse, rat and pig PA28-gamma (proteosome activator 28-gamma, Ki antigen) protein. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction