missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSMD9 Monoclonal Antibody (OTI5H2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527265
This item is not returnable.
View return policy
Description
PSMD9 Monoclonal Antibody for Western Blot
Acts as a chaperone during the assembly of the 26S proteasome, specifically of the base subcomplex of the PA700/19S regulatory complex (RC). During the base subcomplex assembly is part of an intermediate PSMD9:PSMC6:PSMC3 module, also known as modulator trimer complex; PSMD9 is released during the further base assembly process.
Specifications
| PSMD9 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O00233 | |
| PSMD9 | |
| Full length human recombinant protein of PSMD9 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI5H2 | |
| Unconjugated | |
| PSMD9 | |
| 1500011J20Rik; 2610202L11Rik; 26S proteasome non-ATPase regulatory subunit 9; 26S proteasome regulatory subunit p27; Bridge; Bridge-1; homolog of rat Bridge 1; hypothetical protein LOC513315; MGC8644; P27; proteasome (prosome, macropain) 26S subunit, non-ATPase, 9; proteasome 26S non-ATPase regulatory subunit 9; proteasome 26S non-ATPase subunit 9; proteasome 26S subunit, non-ATPase 9; Psmd9; Rpn4; Transactivating protein Bridge-1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5715 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction