missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMD8 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95230-0.1ml
This item is not returnable.
View return policy
Description
PSMD8 Polyclonal antibody specifically detects PSMD8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| PSMD8 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| 26S proteasome non-ATPase regulatory subunit 8, 26S proteasome regulatory subunit p31, HIP6,26S proteasome regulatory subunit S14, HYPF, MGC1660, Nin1p, p3126S proteasome regulatory subunit RPN12, proteasome (prosome, macropain) 26S subunit, non-ATPase, 8, Rpn12, S14 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 101-350 of human PSMD8 (NP_002803.2). EWNRKSPNLSKCGEELGRLKLVLLELNFLPTTGTKLTKQQLILARDILEIGAQWSILRKDIPSFERYMAQLKCYYFDYKEQLPESAYMHQLLGLNLLFLLSQNRVAEFHTELERLPAKDIQTNVYIKHPVSLEQYLMEGSYNKVFLAKGNIPAESYTFFIDILLDTIRDEIAGCIEKAYEKILFTEATRILFFNTPKKMTDYAKKRGWVLGPNNYYSFASQQQKPEDTTIPSTELAKQVIEYARQLEMIV | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 5714 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction