missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMD6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12539-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the protease subunit S10 family. The encoded protein is a subunit of the 26S proteasome which colocalizes with DNA damage foci and is involved in the ATP-dependent degradation of ubiquinated proteins. Alternative splicing results in multiple transcript variants.Specifications
| PSMD6 | |
| Polyclonal | |
| Unconjugated | |
| PSMD6 | |
| KIAA0107, p42A, p44S10, PFAAP4, PSMD6, Rpn7, S10, SGA 113M | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 289924, 66413, 9861 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q15008, Q99JI4 | |
| Psmd6 | |
| PSMD6 Fusion Protein Ag3251 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction