missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSMD6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530333
This item is not returnable.
View return policy
Description
Recommended positive controls: Jurkat, Raji, K562, THP-1. Predicted reactivity: Mouse (98%), Rat (99%), Zebrafish (97%), Xenopus laevis (95%), Rhesus Monkey (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the protease subunit S10 family. The encoded protein is a subunit of the 26S proteasome which colocalizes with DNA damage foci and is involved in the ATP-dependent degradation of ubiquinated proteins. Alternative splicing results in multiple transcript variants.
Specifications
| PSMD6 | |
| Polyclonal | |
| Unconjugated | |
| PSMD6 | |
| 2400006A19Rik; 26S proteasome non-ATPase regulatory subunit 6; 26S proteasome non-ATPase regulatory subunit 6-like protein; 26S proteasome non-ATPase regulatory subunit-like protein; 26S proteasome regulatory subunit RPN7; 26S proteasome regulatory subunit S10; breast cancer-associated protein SGA-113M; KIAA0107; p42A; P44s10; PFAAP4; Phosphonoformate immuno-associated protein 4; proteasome (prosome, macropain) 26S subunit, non-ATPase, 6; proteasome 26S subunit, non-ATPase 6; proteasome regulatory particle subunit p44S10; proteasome, 26S, non-ATPase regulatory subunit 6; PSMD6; Rpn7; S10; SGA-113M; vov; zgc:56471 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 289924, 66413, 9861 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.87 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q15008, Q99JI4 | |
| PSMD6 | |
| Recombinant protein encompassing a sequence within the center region of human PSMD6. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction