missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMD6, Mouse anti-Human, Clone: 1C1, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00009861-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: YRYYVREMRIHAYSQLLESYRSLTLGYMAEAFGVGVEFIDQELSRFIAAGRLHCKIDKVNEIVETNRPDSKNWQYQETIKKGDLLLNRVQKLSRVINMSpecifications
| PSMD6 | |
| Monoclonal | |
| Unconjugated | |
| proteasome (prosome, macropain) 26S subunit, non-ATPase, 6 | |
| KIAA0107/Rpn7/S10/SGA-113M/p44S10 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, KnockDown, Western Blot | |
| 1C1 | |
| In 1x PBS, pH 7.4 | |
| NM_014814 | |
| PSMD6 | |
| PSMD6 (NP_055629, 292 a.a. ∼ 389 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 9861 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction