missing translation for 'onlineSavingsMsg'
Learn More

PSMD6 Antibody, Novus Biologicals™

Product Code. 18397769 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18397769 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18397769 Supplier Novus Biologicals Supplier No. H00009861B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

PSMD6 Polyclonal antibody specifically detects PSMD6 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMD6
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_055629.1
Gene Alias 26S proteasome non-ATPase regulatory subunit 6, Breast cancer-associated protein SGA-113M, KIAA010726S proteasome regulatory subunit S10, p42A, p44S10, PFAAP4, Phosphonoformate immuno-associated protein 4, proteasome (prosome, macropain) 26S subunit, non-ATPase, 6,26S proteasome regulatory subunit RPN7, Proteasome regulatory particle subunit p44S10, Rpn7, S10, SGA-113M
Host Species Mouse
Immunogen PSMD6 (NP_055629.1, 1 a.a. - 389 a.a.) full-length human protein. MPLENLEEEGLPKNPDLRIAQLRFLLSLPEHRGDAAVRDELMAAVRDNNMAPYYEALCKSLDWQIDVDLLNKMKKANEDELKRLDEELEDAEKNLGESEIRDAMMAKAEYLCRIGDKEGALTAFRKTYDKTVALGHRLDIVFYLLRIGLFYMDNDLITRNTEKAKSLIEEGGDWDRRNRLKVYQGLYCVAIRDFKQAAELFLDTVSTFTSYELMDYKTFVTYTVYVSMIALERPDLREKVIKGAEILEVLHSLPAVRQYLFSLYECRYSVFFQSLAVVEQEMKKDWLFAPHYRYYVREMRIHAYSQLLESYRSLTLGYMAEAFGVGVEFIDQELSRFIAAGRLHCKIDKVNEIVETNRPDSKNWQYQETIKKGDLLLNRVQKLSRVINM
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9861
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.